Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ebag9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201094.1 | NP_957388.1 | 2 - 3 | 285 | 73 | |
GenBank mRNA | BC049504.1 | - | 2 - 3 | 285 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |