Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dynlt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205608.1 | NP_991171.1 | 3 - 4 | 282 | 95 | |
GenBank mRNA | AY398423.1 | - | 3 - 4 | 282 | 95 | |
BC081386.1 | - | 3 - 4 | 218 | 95 | ||
BC164919.1 | - | 3 - 4 | 233 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |