Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003744.2 | NP_001003744.1 | 5 | 6536 | 142 | |
NM_001362494.1 | NP_001349423.1 | 6 | 6659 | 142 | ||
NR_155755.1 | 4 | 6357 | 142 | |||
GenBank mRNA | BC078646.1 | - | 6 | 6252 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |