Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cldnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131763.2 | NP_571838.1 | 1 | 72 | 92 | |
GenBank mRNA | AF359426.1 | - | 1 | 47 | 92 | |
BC078358.1 | - | 1 | 51 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Inventoried Both primers and probe map within a single exon |