Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mphosph6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003618.1 | NP_001003618.1 | 4 - 5 | 467 | 109 | |
GenBank mRNA | BC078314.1 | - | 4 - 5 | 467 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |