Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003890.3 | NP_001003890.3 | 8 - 9 | 1523 | 115 | |
GenBank mRNA | AY648752.1 | - | 8 - 9 | 1482 | 115 | |
BC098545.1 | - | 8 - 9 | 1523 | 115 | ||
HM210100.1 | - | 8 - 9 | 1239 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |