Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epha4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005919.2 | NP_001005919.2 | 8 - 9 | 1798 | 150 | |
NM_001365171.1 | NP_001352100.1 | 8 - 9 | 1798 | 150 | ||
XM_005171317.4 | XP_005171374.1 | 8 - 9 | 2080 | 150 | ||
XM_005171318.4 | XP_005171375.1 | 8 - 9 | 2081 | 150 | ||
XM_009298712.3 | XP_009296987.1 | 8 - 9 | 2085 | 150 | ||
GenBank mRNA | BC098529.1 | - | 8 - 9 | 1793 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |