Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oxct1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007291.1 | NP_001007292.1 | 4 - 5 | 447 | 88 | |
GenBank mRNA | BC078408.2 | - | 4 - 5 | 447 | 88 | |
BC163972.1 | - | 4 - 5 | 474 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |