Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhbdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017614.1 | NP_001017614.1 | 6 - 7 | 866 | 97 | |
GenBank mRNA | BC092835.1 | - | 6 - 7 | 866 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |