Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: praf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017698.1 | NP_001017698.1 | 2 - 3 | 394 | 104 | |
GenBank mRNA | BC093309.1 | - | 2 - 3 | 394 | 104 | |
BC164952.1 | - | 2 - 3 | 421 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |