Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b9d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024373.1 | NP_001019544.1 | 1 - 2 | 194 | 105 | |
GenBank mRNA | BC095392.1 | - | 1 - 2 | 194 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |