Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smad6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024810.2 | NP_001019981.2 | 4 | 1780 | 72 | |
GenBank mRNA | AY987387.1 | - | 1 | 441 | 72 | |
BC117577.1 | - | 5 | 670 | 72 | ||
BC128793.1 | - | 4 | 512 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |