Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fdps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025471.1 | NP_001020642.1 | 3 - 4 | 364 | 112 | |
GenBank mRNA | BC083515.1 | - | 2 - 3 | 295 | 112 | |
BC097112.1 | - | 3 - 4 | 364 | 112 | ||
BC164398.1 | - | 2 - 3 | 320 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |