Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: baiap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040245.1 | NP_001035335.1 | 7 - 8 | 739 | 70 | |
XM_005163755.4 | XP_005163812.1 | 7 - 8 | 801 | 70 | ||
XM_009299775.3 | XP_009298050.1 | 7 - 8 | 802 | 70 | ||
XM_009299776.2 | XP_009298051.1 | 7 - 8 | 801 | 70 | ||
XM_021471329.1 | XP_021327004.1 | 7 - 8 | 801 | 70 | ||
XM_021471351.1 | XP_021327026.1 | 7 - 8 | 801 | 70 | ||
GenBank mRNA | BC115281.1 | - | 7 - 8 | 739 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |