Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpusd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076753.1 | NP_001070221.1 | 5 - 6 | 735 | 143 | |
XM_017354591.2 | XP_017210080.1 | 5 - 6 | 819 | 143 | ||
GenBank mRNA | BC124362.1 | - | 5 - 6 | 735 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |