Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c1galt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077374.1 | NP_001070842.1 | 3 - 4 | 1014 | 107 | |
XM_005162255.4 | XP_005162312.1 | 3 - 4 | 1313 | 107 | ||
GenBank mRNA | BC124666.1 | - | 3 - 4 | 1014 | 107 | |
BC164510.1 | - | 2 - 3 | 940 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |