Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2k19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079704.2 | NP_001073172.1 | 4 - 5 | 671 | 71 | |
GenBank mRNA | AY825260.1 | - | 4 - 5 | 672 | 71 | |
BC081666.1 | - | 4 - 5 | 631 | 71 | ||
BC125903.1 | - | 4 - 5 | 656 | 71 | ||
BC171464.1 | - | 4 - 5 | 734 | 71 | ||
BC171468.1 | - | 4 - 5 | 734 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |