Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacng7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083084.1 | NP_001076553.1 | 2 - 3 | 275 | 121 | |
XM_005159404.4 | XP_005159461.1 | 3 - 4 | 604 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |