Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnnc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131563.2 | NP_571638.1 | 3 - 4 | 260 | 83 | |
GenBank mRNA | AF180890.1 | - | 3 - 4 | 257 | 83 | |
BC064284.1 | - | 3 - 4 | 260 | 83 | ||
BC154312.1 | - | 3 - 4 | 274 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |