Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167956.2 | NP_001161428.1 | 5 - 6 | 570 | 107 | |
GenBank mRNA | BC046047.1 | - | 5 - 6 | 482 | 107 | |
EH524379.1 | - | 4 - 5 | 547 | 107 | ||
EH555113.1 | - | 5 - 6 | 614 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |