Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacna1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114548.1 | NP_001108020.1 | 34 - 35 | 4764 | 93 | |
XM_009301497.3 | XP_009299772.1 | 38 - 39 | 5598 | 93 | ||
XM_009301500.3 | XP_009299775.1 | 36 - 37 | 4929 | 93 | ||
XM_009301501.3 | XP_009299776.1 | 37 - 38 | 4899 | 93 | ||
XM_009301504.3 | XP_009299779.1 | 36 - 37 | 4929 | 93 | ||
XM_009301505.3 | XP_009299780.1 | 34 - 35 | 5517 | 93 | ||
XM_017356095.2 | XP_017211584.1 | 38 - 39 | 4935 | 93 | ||
XM_017356096.2 | XP_017211585.1 | 37 - 38 | 4920 | 93 | ||
XM_017356097.2 | XP_017211586.1 | 36 - 37 | 5529 | 93 | ||
XM_021476173.1 | XP_021331848.1 | 38 - 39 | 4962 | 93 | ||
XM_021476174.1 | XP_021331849.1 | 37 - 38 | 4956 | 93 | ||
XM_021476175.1 | XP_021331850.1 | 37 - 38 | 4950 | 93 | ||
XM_021476176.1 | XP_021331851.1 | 37 - 38 | 4941 | 93 | ||
XM_021476178.1 | XP_021331853.1 | 35 - 36 | 4923 | 93 | ||
XM_021476179.1 | XP_021331854.1 | 35 - 36 | 4923 | 93 | ||
XM_021476180.1 | XP_021331855.1 | 38 - 39 | 4962 | 93 | ||
XM_021476181.1 | XP_021331856.1 | 35 - 36 | 5502 | 93 | ||
XM_021476182.1 | XP_021331857.1 | 34 - 35 | 5496 | 93 | ||
GenBank mRNA | KC192784.1 | - | 36 - 37 | 4929 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |