Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vat1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114560.1 | NP_001108032.1 | 2 - 3 | 350 | 81 | |
GenBank mRNA | BC154027.1 | - | 2 - 3 | 350 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |