Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: edar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115064.2 | NP_001108536.2 | 5 - 6 | 575 | 89 | |
GenBank mRNA | EF137867.1 | - | 5 - 6 | 575 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |