Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ewsr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115138.2 | NP_001108610.1 | 12 - 13 | 1391 | 117 | |
XM_021479109.1 | XP_021334784.1 | 12 - 13 | 1390 | 117 | ||
GenBank mRNA | BC056281.1 | - | 12 - 13 | 1368 | 117 | |
BC068397.1 | - | 11 - 12 | 1360 | 117 | ||
DT075886.1 | - | 2 - 3 | 170 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |