Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243037.2 | NP_001229966.1 | 6 - 7 | 551 | 81 | |
NM_001319650.1 | NP_001306579.1 | 6 - 7 | 720 | 81 | ||
GenBank mRNA | BC055097.1 | - | 5 - 6 | 512 | 81 | |
CF996097.1 | - | 5 - 6 | 563 | 81 | ||
EH488885.1 | - | 6 - 7 | 594 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |