Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213015.1 | NP_998180.1 | 2 - 3 | 238 | 69 | |
XM_005157414.4 | XP_005157471.1 | 2 - 3 | 329 | 69 | ||
XM_005157415.4 | XP_005157472.1 | 2 - 3 | 377 | 69 | ||
GenBank mRNA | BC055511.1 | - | 2 - 3 | 270 | 69 | |
BC153596.1 | - | 2 - 3 | 267 | 69 | ||
BC153597.1 | - | 1 - 2 | 244 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |