Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212680.1 | NP_997845.1 | 5 | 1048 | 89 | |
GenBank mRNA | BC065852.1 | - | 5 | 1048 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |