Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109680.1 | NP_001103150.1 | 1 - 2 | 94 | 112 | |
XM_023626553.1 | XP_023482321.1 | 897 | 112 | |||
XM_023626556.1 | XP_023482324.1 | 1618 | 112 | |||
XM_023626563.1 | XP_023482331.1 | 667 | 112 | |||
GenBank mRNA | EU029660.1 | - | 1 - 2 | 94 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |