Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023624431.1 | XP_023480199.1 | 2070 | 103 | ||
GenBank mRNA | AF510665.1 | - | 3 - 4 | 433 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Equus caballus Probe spans exons |