Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163890.2 | NP_001157362.1 | 2 - 3 | 204 | 82 | |
GenBank mRNA | AY246732.1 | - | 1 - 2 | 133 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Equus caballus Probe spans exons |