Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252308.1 | NP_001239237.1 | 2 - 3 | 328 | 67 | |
XM_005609922.3 | XP_005609979.1 | 526 | 67 | |||
XM_005609923.3 | XP_005609980.1 | 526 | 67 | |||
XM_014739697.2 | XP_014595183.1 | 528 | 67 | |||
XM_023640220.1 | XP_023495988.1 | 526 | 67 | |||
XM_023640221.1 | XP_023495989.1 | 529 | 67 | |||
GenBank mRNA | AY376269.1 | - | 1 - 2 | 206 | 67 | |
JF749872.1 | - | 2 - 3 | 328 | 67 | ||
JL640440.1 | - | 2 - 3 | 496 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Equus caballus Probe spans exons |