Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199415.1 | NP_001186344.1 | 4 - 5 | 389 | 101 | |
XM_015286158.2 | XP_015141644.1 | 3 - 4 | 760 | 101 | ||
GenBank mRNA | BU136820.1 | - | 4 - 5 | 392 | 101 | |
BU301058.1 | - | 4 - 5 | 369 | 101 | ||
BX929762.1 | - | 4 - 5 | 323 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |