Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSPRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415546.6 | XP_415546.3 | 5 - 6 | 591 | 67 | |
GenBank mRNA | BX929810.1 | - | 1 - 2 | 46 | 67 | |
CR385441.1 | - | 1 - 2 | 72 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |