Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_418775.5 | XP_418775.3 | 3 - 4 | 225 | 61 | |
GenBank mRNA | AB046393.1 | - | 2 - 3 | 211 | 61 | |
BX930395.1 | - | 2 - 3 | 359 | 61 | ||
BX934909.1 | - | 2 - 3 | 338 | 61 | ||
CR354366.1 | - | 2 - 3 | 779 | 61 |
Gene Symbol: ENTPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004939433.3 | XP_004939490.1 | 12 - 13 | 7593 | 61 | |
XM_015281774.2 | XP_015137260.1 | 12 - 13 | 7582 | 61 | ||
XM_015281775.2 | XP_015137261.1 | 12 - 13 | 4604 | 61 | ||
XM_025147330.1 | XP_025003098.1 | 13 - 14 | 7028 | 61 | ||
GenBank mRNA | CR407270.1 | - | 2 - 3 | 221 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |