Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_421902.6 | XP_421902.2 | 4 - 5 | 600 | 98 | |
GenBank mRNA | BX931186.1 | - | 4 - 5 | 582 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |