Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CISD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420668.6 | XP_420668.1 | 1 - 2 | 169 | 51 | |
GenBank mRNA | BX932115.1 | - | 1 - 2 | 160 | 51 | |
BX932825.2 | - | 1 - 2 | 117 | 51 | ||
BX936123.2 | - | 1 - 2 | 174 | 51 | ||
CR406249.1 | - | 1 - 2 | 163 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |