Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199612.1 | NP_001186541.1 | 2 - 3 | 376 | 90 | |
XM_015281410.2 | XP_015136896.1 | 3 - 4 | 426 | 90 | ||
GenBank mRNA | BX933986.1 | - | 2 - 3 | 250 | 90 | |
BX934713.2 | - | 2 - 3 | 322 | 90 | ||
CN210835.1 | - | 2 - 3 | 376 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |