Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277465.1 | NP_001264394.1 | 1 - 2 | 111 | 79 | |
NM_001277468.1 | NP_001264397.1 | 1 - 2 | 111 | 79 | ||
GenBank mRNA | BX931358.2 | - | 1 - 2 | 103 | 79 | |
BX933533.2 | - | 1 - 2 | 77 | 79 | ||
CR353858.1 | - | 1 - 2 | 102 | 79 | ||
CR386803.1 | - | 1 - 2 | 89 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |