Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277670.1 | NP_001264599.1 | 2 - 3 | 996 | 62 | |
XM_015278290.2 | XP_015133776.1 | 4 - 5 | 4417 | 62 | ||
XM_025149569.1 | XP_025005337.1 | 4 - 5 | 4246 | 62 | ||
XM_025149570.1 | XP_025005338.1 | 5 - 6 | 4330 | 62 | ||
XM_025149571.1 | XP_025005339.1 | 5 - 6 | 2919 | 62 | ||
XM_025149572.1 | XP_025005340.1 | 5 - 6 | 2425 | 62 | ||
GenBank mRNA | BX933547.2 | - | 2 - 3 | 956 | 62 | |
CR354354.1 | - | 2 - 3 | 1008 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |