Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277376.1 | NP_001264305.1 | 3 - 4 | 283 | 64 | |
GenBank mRNA | BU114844.1 | - | 4 - 5 | 375 | 64 | |
CR524287.1 | - | 4 - 5 | 375 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |