Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198668.1 | NP_001185597.1 | 3 - 4 | 479 | 141 | |
GenBank mRNA | BU109195.1 | - | 3 - 4 | 479 | 141 | |
M21275.1 | - | 3 - 4 | 313 | 141 | ||
U25823.1 | - | 3 - 4 | 306 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |