Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199715.2 | NP_001186644.2 | 1 - 2 | 276 | 245 | |
GenBank mRNA | BX950827.2 | - | 1 - 2 | 180 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |