Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXNAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199638.1 | NP_001186567.1 | 4 - 5 | 331 | 73 | |
XM_015281479.2 | XP_015136965.1 | 4 - 5 | 384 | 73 | ||
XM_015281480.2 | XP_015136966.1 | 4 - 5 | 450 | 73 | ||
XM_015281481.2 | XP_015136967.1 | 4 - 5 | 352 | 73 | ||
XM_015281482.2 | XP_015136968.1 | 3 - 4 | 255 | 73 | ||
GenBank mRNA | BU134990.1 | - | 4 - 5 | 340 | 73 | |
BX932544.2 | - | 3 - 4 | 553 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |