Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_417105.6 | XP_417105.2 | 1 - 2 | 815 | 55 | |
GenBank mRNA | BX929798.2 | - | 1 - 2 | 582 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |