Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277412.1 | NP_001264341.1 | 1 - 2 | 185 | 144 | |
GenBank mRNA | BU104977.1 | - | 1 - 2 | 173 | 144 | |
BU245000.1 | - | 1 - 2 | 181 | 144 | ||
BX930716.1 | - | 1 - 2 | 137 | 144 | ||
BX932323.2 | - | 1 - 2 | 196 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |