Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277443.1 | NP_001264372.1 | 2 - 3 | 256 | 125 | |
GenBank mRNA | BU122238.1 | - | 2 - 3 | 254 | 125 | |
CR390124.1 | - | 2 - 3 | 237 | 125 | ||
CV857335.1 | - | 2 - 3 | 263 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |