Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_424383.6 | XP_424383.4 | 3 - 4 | 563 | 80 | |
GenBank mRNA | CR407322.1 | - | 1 - 2 | 110 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |