Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BECN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006332.1 | NP_001006332.1 | 3 - 4 | 312 | 56 | |
XM_015299595.2 | XP_015155081.1 | 3 - 4 | 365 | 56 | ||
GenBank mRNA | AJ720008.1 | - | 3 - 4 | 312 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |