Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204584.1 | NP_989915.1 | 2 - 3 | 167 | 127 | |
GenBank mRNA | X64539.1 | - | 167 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |