Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204823.1 | NP_990154.1 | 1 - 2 | 572 | 92 | |
GenBank mRNA | AF139721.1 | - | 1 - 2 | 526 | 92 | |
AF179484.1 | - | 1 - 2 | 572 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |